Tisk…

Bis(tri-tert-butylphosphine)palladium(0), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

676578-1G 676578-250MG
SIAL676578-1GEA 12240 CZK
SIAL676578-1G SIAL676578-250MG
Bis(tri-tert-butylphosphine)palladium(0), Sigma-Aldrich®
Bis(tri-tert-butylphosphine)palladium(0)
Vzorec: C₂₄H₅₄P₂Pd
MDL Number: MFCD03094580
CAS číslo: 53199-31-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR