Tisk…

2,7-Di-tert-butylfluorene, Sigma-Aldrich®

Dodavatel: Merck

543306-25G 543306-5G
SIAL543306-25GEA 17680 CZK
SIAL543306-25G SIAL543306-5G
2,7-Di-tert-butylfluorene, Sigma-Aldrich®
2,7-Di-tert-butylfluorene
Vzorec: C₂₁H₂₆
MW: 278,43 g/mol
MDL Number: MFCD03093998
CAS číslo: 58775-05-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR