2,7-Di-tert-butylfluorene ≥97%
Dodavatel: Thermo Fisher Scientific
H55430.06
H55430.14
H55430.06EA
3360
CZK
H55430.06
H55430.14
2,7-Di-tert-butylfluorene ≥97%
2,7-Di-tert-butylfluorene
Purity: 98%
CAS No.: 58775-05-6
Molecular Formula: C21H26
Formula Weight: 278.43
Melting Point: 121-124°
MDL No.: MFCD03093998
Vzorec:
C₂₁H₂₆ Bod tání: 121…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD03093998 CAS číslo: 58775-05-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...