2,6-Diacetylpyridine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
D8801-25G
D8801-1G
D8801-5G
SIALD8801-25GEA
16320
CZK
SIALD8801-25G
SIALD8801-1G
SIALD8801-5G
2,6-Diacetylpyridine, Sigma-Aldrich®
2,6-Diacetylpyridine
Vzorec:
C₉H₉NO₂ |
MDL Number:
MFCD00006304 CAS číslo: 1129-30-2 EINECS: 214-442-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...