Sodium nicotinate ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
B22224.22
B22224.36
B22224.22EA
1390
CZK
B22224.22
B22224.36
Sodium nicotinate ≥98%
Sodium nicotinate
Purity: 98%
CAS No.: 54-86-4
Molecular Formula: C6H4NNaO2
Formula Weight: 145.10
Melting Point:
Vzorec:
C₆H₄NNaO₂ MW: 145,10 g/mol Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064352 CAS číslo: 54-86-4 EINECS: 200-215-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...