Tisk…

Dimethyl-2,6-pyridinedicarboxylate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Dimethyl pyridine-2,6-dicarboxylate

H32251.18 H32251.09
H32251.18EA 5050 CZK
H32251.18 H32251.09
Dimethyl-2,6-pyridinedicarboxylate ≥98%
Dimethyl-2,6-pyridinedicarboxylate

Purity: 98% CAS No.: 5453-67-8 Molecular Formula: C9H9NO4 Formula Weight: 195.17 Melting Point: 119-122° MDL No.: MFCD00134493

Vzorec: C₉H₉NO₄
Bod tání: 119…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00134493
CAS číslo: 5453-67-8
EINECS: 226-697-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR