Tisk…

2,6-Dimethoxy-3-pyridineboronic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2,6-Dimethoxypyridine-3-boronic acid, 2,6-Dimethoxypyridin-3-ylboronic acid

M-Clarity™ quality level = MQ100

706086-1G 706086-250MG
SIAL706086-1GEA 3620 CZK
SIAL706086-1G SIAL706086-250MG
2,6-Dimethoxy-3-pyridineboronic acid, Sigma-Aldrich®
2,6-Dimethoxy-3-pyridineboronic acid
Vzorec: C₇H₁₀BNO₄
MW: 182,97 g/mol
MDL Number: MFCD03788239
CAS číslo: 221006-70-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR