Tisk…

2-Methoxy-3-pyridineboronic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Methoxypyridine-3-boronic acid

M-Clarity™ quality level = MQ100

684589-1G 684589-5G
SIAL684589-1GEA 2100 CZK
SIAL684589-1G SIAL684589-5G
2-Methoxy-3-pyridineboronic acid, Sigma-Aldrich®
2-Methoxy-3-pyridineboronic acid
Vzorec: C₆H₈BNO₃
MW: 152,95 g/mol
MDL Number: MFCD03411572
CAS číslo: 163105-90-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR