Tisk…

Cyclopentanesulphonyl chloride ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H32865.MD H32865.03 H32865.06
H32865.MDEA 2500 CZK
H32865.MD H32865.03 H32865.06
Cyclopentanesulphonyl chloride ≥97%
Cyclopentanesulphonyl chloride

Purity: 97% CAS No.: 26394-17-2 Molecular Formula: C5H9ClO2S Formula Weight: 168.64 Boiling Point: 71-73°/2mm UN No.: 3265 MDL No.: MFCD04117774

Vzorec: C₅H₉ClO₂S
Bod varu: 71…73 °C (2 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD04117774
CAS číslo: 26394-17-2
UN: 3265
ADR: 8,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR