Tisk…

4-(4-Fluorophenoxy)benzaldehyde ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

H50452.03 H50452.MD
H50452.03EA 3320 CZK
H50452.03 H50452.MD
4-(4-Fluorophenoxy)benzaldehyde ≥97%
4-(4-Fluorophenoxy)benzaldehyde

Purity: 97% CAS No.: 137736-06-2 Molecular Formula: C13H9FO2 Formula Weight: 216.21 MDL No.: MFCD01631896

Vzorec: C₁₃H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD01631896
CAS číslo: 137736-06-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR