Tisk…

2-(4-Chlorophenoxy)-6-fluorobenzaldehyde ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

H50847.03 H50847.MD
H50847.03EA 7970 CZK
H50847.03 H50847.MD
2-(4-Chlorophenoxy)-6-fluorobenzaldehyde ≥98%
2-(4-Chlorophenoxy)-6-fluorobenzaldehyde

Purity: 98% CAS No.: 902836-82-2 Molecular Formula: C13H8ClFO2 Formula Weight: 250.66 MDL No.: MFCD08061024

Vzorec: C₁₃H₈ClFO₂
MW: 250,66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061024
CAS číslo: 902836-82-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR