Tisk…

2-Acetoxyisobutyroyl chloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

326178-25G 326178-5G
SIAL326178-25GEA 5780 CZK
SIAL326178-25G SIAL326178-5G
2-Acetoxyisobutyroyl chloride, Sigma-Aldrich®
2-Acetoxyisobutyroyl chloride
Vzorec: C₆H₉ClO₃
MDL Number: MFCD00000708
CAS číslo: 40635-66-3
EINECS: 255-016-2
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR