2-Acetoxyisobutyroyl bromide 96%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Acetoxy-2-methylpropionyl bromide
L11592.14
L11592.22
L11592.14EA
2370
CZK
L11592.14
L11592.22
2-Acetoxyisobutyroyl bromide 96%
2-Acetoxyisobutyroyl bromide
Purity: 96%
CAS No.: 40635-67-4
Molecular Formula: C6H9BrO3
Formula Weight: 209.05
Storage Temperature: 2°C to 8°C
Flash Point:
Vzorec:
C₆H₉BrO₃ MW: 209,05 g/mol Bod varu: 100…110 °C (25 mmHg) Hustota: 1,420 Bod vzplanutí: >110 °C (230 °F) |
MDL Number:
MFCD00040923 CAS číslo: 40635-67-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...