Tisk…

2-Bromoisobutyryl bromide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Bromo-2-methylpropionyl bromide

M-Clarity™ quality level = MQ100

252271-100G 252271-500G
SIAL252271-100GEA 1620 CZK
SIAL252271-100G SIAL252271-500G
2-Bromoisobutyryl bromide, Sigma-Aldrich®
2-Bromoisobutyryl bromide
Vzorec: C₄H₆Br₂O
MDL Number: MFCD00000122
CAS číslo: 20769-85-1
EINECS: 244-017-3
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR