Hypotaurine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Aminoethane-1-sulphinic acid
H1384-100MG
H1384-10MG
H1384-250MG
H1384-1G
SIALH1384-100MGEA
1570
CZK
SIALH1384-100MG
SIALH1384-10MG
SIALH1384-250MG
SIALH1384-1G
Hypotaurine, Sigma-Aldrich®
Hypotaurine
Vzorec:
C₂H₇NO₂S MW: 109.147 g/mol |
MDL Number:
MFCD00038197 CAS číslo: 300-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...