N-Arachidonoyl taurine
Dodavatel: Cayman Chemical
10005537-1
10005537-10
10005537-5
10005537-500
CAYM10005537-1EA
3310
CZK
CAYM10005537-1
CAYM10005537-10
CAYM10005537-5
CAYM10005537-500
N-Arachidonoyl taurine
N-Arachidonoyl taurine
Vzorec:
C₂₂H₃₇NO₄S |
CAS číslo:
119959-65-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...