N-Palmitoyl taurine
Dodavatel: Cayman Chemical
10005611-1
10005611-10
10005611-5
CAYM10005611-1EA
3040
CZK
CAYM10005611-1
CAYM10005611-10
CAYM10005611-5
N-Palmitoyl taurine
N-Palmitoyl taurine
Vzorec:
C₁₈H₃₇NO₄S |
CAS číslo:
83982-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...