Tisk…

3-Iodochromone ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

H34295.MD H34295.03
H34295.MDEA 3570 CZK
H34295.MD H34295.03
3-Iodochromone ≥97%
3-Iodochromone
Vzorec: C₉H₅IO₂
MW: 272,04 g/mol
Bod tání: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD09475514
CAS číslo: 122775-34-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR