Tisk…

3-Bromochromone ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 3-Bromo-4H-chromen-4-one

A14591.03 A14591.06
A14591.03EA 1510 CZK
A14591.03 A14591.06
3-Bromochromone ≥97%
3-Bromochromone

Purity: 97% CAS No.: 49619-82-1 Molecular Formula: C9H5BrO2 Formula Weight: 225.05 Melting Point: 94-98° Beilstein Reference: 125684

Vzorec: C₉H₅BrO₂
MW: 225.05 g/mol
Bod tání: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00017337
CAS číslo: 49619-82-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR