Tisk…
Capillary columns, OPTIMA® 5 MS
Dodavatel: MACHEREY-NAGEL
726210.12 726210.25 726210.50 726211.30 726212.25 726212.50 726212.60 726213.30 726215.12 726215.25 726215.50 726220.15 726220.30 726220.60 726225.30 726225.60 726226.30 726226.60
MANA726210.12EA 11330 CZK
MANA726210.12 MANA726210.25 MANA726210.50 MANA726211.30 MANA726212.25 MANA726212.50 MANA726212.60 MANA726213.30 MANA726215.12 MANA726215.25 MANA726215.50 MANA726220.15 MANA726220.30 MANA726220.60 MANA726225.30 MANA726225.60 MANA726226.30 MANA726226.60
Capillary columns, OPTIMA® 5 MS
Kolony chromatografické
Nonpolar GC phase with low bleeding.
  • Suited for GC/MS and ECD applications and general analyses at trace level
  • Maximum temperature for isothermal operation 340 °C
  • Maximum temperature for short isotherms in a temperature program 360 °C
5% diphenyl –95% dimethylpolysiloxane phase.
USP code: G27/G36.