Tisk…
Black filter paper for the detection of light precipitates, grade MN 220
Dodavatel: MACHEREY-NAGEL
409007 409005 409009 409011 409012 409015 409018
MANA409007EA 509 CZK
MANA409007 MANA409005 MANA409009 MANA409011 MANA409012 MANA409015 MANA409018
Black filter paper for the detection of light precipitates, grade MN 220
Filtry Papíry filtrační Papír kvalitativní filtrační
This filter paper, which is dyed black with a sulphur dye, is used to identify small quantities of light precipitates. For example, it is used for the detection of fluorine or silicon.
Objednat
Produktová specifikace
Environmentally Preferable
Hmotnost 85 g/m²
Tloušťka 0,17 mm
Rychlost filtrace 45 s