5-Azacytosine, (max. 7% H₂O) 98% (dry weight)
Dodavatel: Thermo Scientific
Synonyms:
4-Amino-1,3,5-triazin-2(1H)-one, 6-Amino-1,3,5-triazin-2(1H)-one, 2-Amino-4-hydroxy-1,3,5-triazine
A13410.06
A13410.14
A13410.06EA
1310
CZK
A13410.06
A13410.14
5-Azacytosine, (max. 7% H₂O) 98% (dry weight)
5-Azacytosine
Purity: 98%
CAS No.: 931-86-2
Molecular Formula: C3H4N4O
Formula Weight: 112.09
Melting Point:
Vzorec:
C₃H₄N₄O MW: 112,09 g/mol Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149402 CAS číslo: 931-86-2 EINECS: 213-242-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...