Etidronate disodium hydrate ≥97% (by NMR), Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ100
P5248-100MG
P5248-10MG
SIALP5248-100MGEA
18500
CZK
SIALP5248-100MG
SIALP5248-10MG
Etidronate disodium hydrate ≥97% (by NMR), Sigma-Aldrich®
Etidronate disodium hydrate
Vzorec:
C₂H₆Na₂O₇P₂·H₂O Bod tání: > 300 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00152567 CAS číslo: 7414-83-7 EINECS: 231-025-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...