N-Fmoc-L-leucine ≥98%
Dodavatel: Thermo Scientific
B21040.06
B21040.14
B21040.06EA
1310
CZK
B21040.06
B21040.14
N-Fmoc-L-leucine ≥98%
N-Fmoc-L-leucine
Purity: 98%
CAS No.: 35661-60-0
Molecular Formula: C21H23NO4
Formula Weight: 353.42
Storage Temperature: 2°C to 8°C
Melting Point: 152-155°
Beilstein Reference: 2178254
Optical Rotation: -25° (c=1 in DMF)
MDL No.: MFCD00037133
Vzorec:
C₂₁H₂₃NO₄ MW: 353,4 g/mol Bod tání: 152…155 °C |
MDL Number:
MFCD00037133 CAS číslo: 35661-60-0 EINECS: 252-662-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...