Tisk…

(±)-α-Lipoic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)​-​1,2-​Dithiolane-​3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid

M-Clarity™ quality level = MQ200

62320-5G-F 62320-25G-F
SIAL62320-5G-FEA 2460 CZK
SIAL62320-5G-F SIAL62320-25G-F
(±)-α-Lipoic acid, Sigma-Aldrich®
(±)-α-Lipoic acid
Vzorec: C₈H₁₄O₂S₂
MDL Number: MFCD00005474
CAS číslo: 1077-28-7
EINECS: 214-071-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR