Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate ≥98%
Dodavatel: abcr
|
Varování
|
Synonyms:
Bis(1,5-cyclooctadiene)iridium (I) tetrafluoroborate
AB120646.0002
AB120646.0500
ABCRAB120646.0002EA
0
CZK
ABCRAB120646.0002
ABCRAB120646.0500
Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate ≥98%
Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate
Vzorec:
C₁₆H₂₄BF₄Ir |
MDL Number:
MFCD00079664 CAS číslo: 35138-23-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...