1-Phenylcyclohexanecarboxylic acid 95%
Dodavatel: Thermo Fisher Scientific
Synonyms:
1-Phenyl-1-cyclohexanecarboxylic acid
170750100
170750500
ACRO170750100EA
2210
CZK
ACRO170750100
ACRO170750500
1-Phenylcyclohexanecarboxylic acid 95%
1-Phenylcyclohexanecarboxylic acid
Vzorec:
C₁₃H₁₆O₂ Bod tání: 119…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037152 CAS číslo: 1135-67-7 EINECS: 214-495-8 |
Výsledky specifikačního testu
Appearance | Beige to brown Powder |
Infrared spectrum | Conforms |
Melting point | 119 to 124 °C |
Titration with NaOH | ≥94 % |
Stabilizer | Approx. 200 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...