Tisk…

Methyl-1-cyclopentene-1-carboxylate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 1-cyclopentene-1-carboxylate

422436-10G 422436-1G
SIAL422436-10GEA 0 CZK
SIAL422436-10G SIAL422436-1G
Methyl-1-cyclopentene-1-carboxylate, Sigma-Aldrich®
Methyl-1-cyclopentene-1-carboxylate
Vzorec: C₇H₁₀O₂
MW: 126,1531 g/mol
MDL Number: MFCD00239506
CAS číslo: 25662-28-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR