Tisk…

1-(Trifluoromethyl)cyclopentanecarboxylic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

H26055.MD H26055.03
H26055.MDEA 3990 CZK
H26055.MD H26055.03
1-(Trifluoromethyl)cyclopentanecarboxylic acid ≥97%
1-(Trifluoromethyl)cyclopentanecarboxylic acid

Purity: 97% CAS No.: 277756-44-2 Molecular Formula: C7H9F3O2 Formula Weight: 182.15 Melting Point: 36-38° MDL No.: MFCD08445821

Vzorec: C₇H₉F₃O₂
Bod tání: 36…38 °C
Storage Temperature: Ambient
MDL Number: MFCD08445821
CAS číslo: 277756-44-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR