Methyl cyclohexanecarboxylate 98%
Dodavatel: Thermo Scientific
|
Varování
|
188820500
188820050
ACRO188820500EA
3340
CZK
ACRO188820500
ACRO188820050
Methyl cyclohexanecarboxylate 98%
Methyl cyclohexanecarboxylate
Vzorec:
C₈H₁₄O₂ MW: 142,2 g/mol Hustota: 0,995 g/cm³ Bod vzplanutí: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001458 CAS číslo: 4630-82-4 EINECS: 225-050-2 UN: 3272 ADR: 3,III |
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4420 to 1.4440 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...