4'-(Methylthio)acetophenone ≥99%
Dodavatel: Thermo Scientific
Synonyms:
1-(4-(Methylthio)phenyl)ethanone
L18074.14
L18074.06
L18074.14EA
9610
CZK
L18074.14
L18074.06
4'-(Methylthio)acetophenone ≥99%
4'-(Methylthio)acetophenone
Purity: 99%
CAS No.: 1778-09-2
Molecular Formula: C9H10OS
Formula Weight: 166.24
Melting Point: 80-82°
Boiling Point: 135°/1mm
MDL No.: MFCD00039835
Vzorec:
C₉H₁₀OS MW: 166,24 g/mol Bod varu: 135 °C (1 mmHg) Bod tání: 80…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039835 CAS číslo: 1778-09-2 EINECS: 217-213-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...