Tisk…

2-Aminoacetophenone hydrochloride 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Phenacylamine hydrochloride

H29117.03 H29117.09
H29117.03EA 1460 CZK
H29117.03 H29117.09
2-Aminoacetophenone hydrochloride 97%
2-Aminoacetophenone hydrochloride

Purity: 98% CAS No.: 5468-37-1 Molecular Formula: C8H9NO.HCl Formula Weight: 171.63 Odour: Odourless Melting Point: ca 194° dec. MDL No.: MFCD00012873

Vzorec: C₈H₁₀ClNO
MW: 171.63 g/mol
Bod tání: ∼ 194 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00012873
CAS číslo: 5468-37-1
EINECS: 226-787-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR