Tisk…

Monomethyl terephthalate 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Methyl hydrogen terephthalate, 4-(Methoxycarbonyl)benzoic acid

352340050 352341000 352340250
ACRO352340050EA 1410 CZK
ACRO352340050 ACRO352341000 ACRO352340250
Monomethyl terephthalate 97%
Monomethyl terephthalate
Vzorec: C₉H₈O₄
Bod tání: 219…224 °C
Storage Temperature: Ambient
MDL Number: MFCD00002557
CAS číslo: 1679-64-7
EINECS: 216-849-7

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 219 to 224 °C
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR