Tisk…

DSS (Disuccinimidyl suberate), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: DSS

M-Clarity™ quality level = MQ200

S1885-1G S1885-500MG S1885-5G
SIALS1885-1GEA 4230 CZK
SIALS1885-1G SIALS1885-500MG SIALS1885-5G
DSS (Disuccinimidyl suberate), Sigma-Aldrich®
DSS (Disuccinimidyl suberate)
Vzorec: C₁₆H₂₀N₂O₈
MDL Number: MFCD00049059
CAS číslo: 68528-80-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR