Tisk…

N-Acetyl-L-leucine 99%

Dodavatel: Thermo Scientific

Synonyms: N-Ac-Leu-OH, Ac-Leu-OH

347010050 347010250
ACRO347010050EA 1310 CZK
ACRO347010050 ACRO347010250
N-Acetyl-L-leucine 99%
N-Acetyl-L-leucine
Vzorec: C₈H₁₅NO₃
MW: 173,2 g/mol
Bod tání: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00065131
CAS číslo: 1188-21-2
EINECS: 214-706-3

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder
Infrared spectrum Conforms
Solubility (5 % in ethanol) Clear colorless
Specific optical rotation -23° to -26° (20°C, 589 nm) (c=4, MeOH)
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR