Tisk…
Dimethyl cyclohexane-1,4-dicarboxylate (mixture of cis and trans isomers), Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Dimethyl cyclohexane-1,4-dicarboxylate
206431-100G 206431-25G
SIAL206431-100GEA 6890 CZK
SIAL206431-100G SIAL206431-25G
Dimethyl cyclohexane-1,4-dicarboxylate (mixture of cis and trans isomers), Sigma-Aldrich®
Dimethyl cyclohexane-1,4-dicarboxylate (mixture of cis and trans isomers)
Vzorec: C₁₀H₁₆O₄
MW: 200.2316 g/mol
MDL Number: MFCD00001460
CAS číslo: 94-60-0
EINECS: 202-347-5