2,4,5,6-Tetraaminopyrimidine sulphate 97%
Dodavatel: Thermo Scientific
|
Varování
|
A18788.09
A18788.18
A18788.30
A18788.09EA
975
CZK
A18788.09
A18788.18
A18788.30
2,4,5,6-Tetraaminopyrimidine sulphate 97%
2,4,5,6-Tetraaminopyrimidine sulphate
Used as an intermediate in the production of Methotrexate.
Purity: 97%
CAS No.: 5392-28-9
Molecular Formula: C4H8N6.H2SO4
Formula Weight: 238.23
Melting Point:
Vzorec:
C₄H₈N₆·H₂SO₄ MW: 238,23 g/mol Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012786 CAS číslo: 5392-28-9 EINECS: 226-393-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...