3-(3,4-Dihydroxyphenyl)-L-alanine ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Dopar, L-dopa, L-β-3,4-Dihydroxyphenyl-Alanine, 3-Hydroxy-L-tyrosine, Levodopa
A11311.06
A11311.14
A11311.22
A11311.06EA
1380
CZK
A11311.06
A11311.14
A11311.22
3-(3,4-Dihydroxyphenyl)-L-alanine ≥98%
3-(3,4-Dihydroxyphenyl)-L-alanine
Purity: 98+%
CAS No.: 59-92-7
Molecular Formula: C9H11NO4
Formula Weight: 197.19
Odour: Odourless
Melting Point: ca 295° dec.
Beilstein Reference: 2215169
Merck Reference: 14,5464
MDL No.: MFCD00002598
Vzorec:
C₉H₁₁NO₄ MW: 197,19 g/mol Bod tání: ∼ 295 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00002598 CAS číslo: 59-92-7 EINECS: 200-445-2 Merck Index: 14,5464 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...