2-Chloro-L-phenylalanine ≥98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
H-Phe(2-Cl)-OH
H27135.MD
H27135.03
H27135.MDEA
5010
CZK
H27135.MD
H27135.03
2-Chloro-L-phenylalanine ≥98%
2-Chloro-L-phenylalanine
Purity: 98+%
CAS No.: 103616-89-3
Molecular Formula: C9H10ClNO2
Formula Weight: 199.64
Melting Point: 196-199°
MDL No.: MFCD00077921
Vzorec:
C₉H₁₀ClNO₂ MW: 199.6 g/mol Bod tání: 196…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077921 CAS číslo: 103616-89-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...