Tisk…

4-Chloro-D-phenylalanine ≥95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: H-D-Phe(4-Cl)-OH, (R)-2-Amino-3-(4-chlorophenyl)propionic acid, H-p-Chloro-D-Phe-OH

H51982.MD H51982.03 H51982.06
H51982.MDEA 1550 CZK
H51982.MD H51982.03 H51982.06
4-Chloro-D-phenylalanine ≥95%
4-Chloro-D-phenylalanine

Purity: 95% CAS No.: 14091-08-8 Molecular Formula: C9H10ClNO2 Formula Weight: 199.64 Melting Point: 260° Beilstein Reference: 2416151 UN No.: 2811 MDL No.: MFCD00079675

Vzorec: C₉H₁₀ClNO₂
MW: 199.64 g/mol
Bod tání: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00079675
CAS číslo: 14091-08-8
UN: 2811
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR