Tisk…

4-Fluoro-DL-phenylalanine ≥98%

Dodavatel: Thermo Scientific

L07585.03 L07585.06
L07585.03EA 1420 CZK
L07585.03 L07585.06
4-Fluoro-DL-phenylalanine ≥98%
4-Fluoro-DL-phenylalanine

Purity: 98+% CAS No.: 51-65-0 Molecular Formula: C9H10FNO2 Formula Weight: 183.18 Melting Point: ca 254° dec. Beilstein Reference: 29338793

Vzorec: C₉H₁₀FNO₂
MW: 183,18 g/mol
Bod tání: ∼ 254 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00002600
CAS číslo: 51-65-0
EINECS: 200-113-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR