Tisk…

4-Bromo-DL-phenylalanine ≥98%

Dodavatel: Thermo Fisher Scientific

L02833.03 L02833.06
L02833.03EA 1930 CZK
L02833.03 L02833.06
4-Bromo-DL-phenylalanine ≥98%
4-Bromo-DL-phenylalanine

Purity: 98+% CAS No.: 14091-15-7 Molecular Formula: C9H10BrNO2 Formula Weight: 244.09 Odour: Odourless Melting Point: ca 262° dec. Beilstein Reference: 2212158 MDL No.: MFCD00002599

Vzorec: C₉H₁₀BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD00002599
CAS číslo: 14091-15-7
EINECS: 237-941-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR