Tisk…

1-(3-Aminopropyl)pyrrolidine 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L04739.06 L04739.14
L04739.06EA 2270 CZK
L04739.06 L04739.14
1-(3-Aminopropyl)pyrrolidine 97%
1-(3-Aminopropyl)pyrrolidine

Purity: 97% CAS No.: 23159-07-1 Molecular Formula: C7H16N2 Formula Weight: 128.22 Refractive Index: 1.4710 Boiling Point: 191-193° UN No.: 2735 MDL No.: MFCD00014100

Vzorec: C₇H₁₆N₂
Bod varu: 191…193 °C
Hustota: 1,014
Storage Temperature: Ambient
MDL Number: MFCD00014100
CAS číslo: 23159-07-1
EINECS: 245-465-2
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR