Tisk…

trans-4-Aminocyclohexanecarboxylic acid 97%

Dodavatel: Thermo Scientific

H27489.03 H27489.06
H27489.03EA 3330 CZK
H27489.03 H27489.06
trans-4-Aminocyclohexanecarboxylic acid 97%
trans-4-Aminocyclohexanecarboxylic acid

Purity: 97% CAS No.: 3685-25-4 Molecular Formula: C7H13NO2 Formula Weight: 143.19 Melting Point: 300° MDL No.: MFCD00209953

Vzorec: C₇H₁₃NO₂
MW: 143,19 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00209953
CAS číslo: 3685-25-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR