Tisk…
Sodium decanoate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
C4151-100G C4151-5G
SIALC4151-100GEA 27490 CZK
SIALC4151-100G SIALC4151-5G
Sodium decanoate, Sigma-Aldrich®
Sodium decanoate
Vzorec: C₁₀H₁₉NaO₂
MDL Number: MFCD00066453
CAS číslo: 1002-62-6
EINECS: 213-688-4