Gellan gum, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
P8169-100G
P8169-1KG
P8169-250G
P8169-500G
P8169-5KG
SIALP8169-100GEA
1730
CZK
SIALP8169-100G
SIALP8169-1KG
SIALP8169-250G
SIALP8169-500G
SIALP8169-5KG
Gellan gum, Sigma-Aldrich®
Gellan gum
Storage Temperature:
Ambient |
MDL Number:
MFCD00131909 CAS číslo: 71010-52-1 EINECS: 275-117-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...