Tisk…

α-Bromostyrene, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: alfa-Bromostyrene, α-Bromostyrene, alpha-Bromostyrene

M-Clarity™ quality level = MQ100

292273-25G 292273-5G
SIAL292273-25GEA 12280 CZK
SIAL292273-25G SIAL292273-5G
α-Bromostyrene, Sigma-Aldrich®
α-Bromostyrene
Vzorec: C₈H₇Br
MW: 183,0452 g/mol
MDL Number: MFCD00012229
CAS číslo: 98-81-7
EINECS: 202-702-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR