Tisk…

Monomethyl succinate 95%

Dodavatel: Thermo Scientific

Synonyms: 4-Methoxy-4-oxobutanoic acid, Methyl hydrogen succinate, Succinic acid monomethyl ester

127730100 127730250 127731000
ACRO127730100EA 1060 CZK
ACRO127730100 ACRO127730250 ACRO127731000
Monomethyl succinate 95%
Monomethyl succinate
Vzorec: C₅H₈O₄
MW: 132,11 g/mol
Bod varu: 151 °C (20 mmHg)
Bod tání: 55…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00002788
CAS číslo: 3878-55-5
EINECS: 223-408-2

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystals and/or chunks
Infrared spectrum Conforms
Melting point 52 to 58 °C
GC ≥94 %
Sodium oxide (Na2O) ≤0.4 %
Silicon dioxide (SiO2) ≤0.03 %
Iron(III)oxide (Fe2O3) ≤0.03 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR