Tisk…

D-tert-Leucine (H-D-Tle-OH) ≥99%

Dodavatel: Thermo Fisher Scientific

Synonyms: H-D-Tle-OH

L14098.04 L14098.ME
L14098.04EA 17810 CZK
L14098.04 L14098.ME
D-tert-Leucine (H-D-Tle-OH) ≥99%
D-tert-Leucine (H-D-Tle-OH)

Purity: 99% CAS No.: 26782-71-8 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: 300° Beilstein Reference: 1721825

Vzorec: C₆H₁₃NO₂
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004265
CAS číslo: 26782-71-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR