Tisk…

Isopropyl-4-aminobenzoate 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Isopropyl 4-aminobenzoate

A17551.14 A17551.22
A17551.14EA 1610 CZK
A17551.14 A17551.22
Isopropyl-4-aminobenzoate 98%
Isopropyl-4-aminobenzoate

Purity: 98% CAS No.: 18144-43-9 Molecular Formula: C10H13NO2 Formula Weight: 179.21 MDL No.: MFCD00017108

Vzorec: C₁₀H₁₃NO₂
MW: 179,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017108
CAS číslo: 18144-43-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR